Order Entry
Poland
ContactUsLinkComponent
2-Iodophenyl isothiocyanate ≥97%
Toxic
Corrosive
2-Iodophenyl isothiocyanate ≥97%
Catalog # L11012.03
CAS Number:  
Toxic
Corrosive
2-Iodophenyl isothiocyanate ≥97%
Catalog # L11012.03
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

CAS Number:98041-44-2
 :
Formula:C₇H₄INS
MDL Number:MFCD00041057
 :261.09 g/mol
Boiling Pt:296 °C (1013 hPa)
Melting Pt:36…39 °C
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
UN:2923
ADR:8,III
 :GHS06
 :GHS05