DFDNB (1,5-Difluoro-2,4-dinitrobenzene), Sigma-Aldrich®
Catalog # SIALD102504-10G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:204.09 g/mol
Boiling Pt:132 °C (2 mmHg)
Melting Pt:74…76 °C
Density:1.86 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:206-324-0
UN:2811
ADR:6.1,II
:M-Clarity™ quality level = MQ100
CAS Number:327-92-4
Formula:C₆H₂F₂N₂O₄
MDL Number:MFCD00007052