4-Fluorophenyl isothiocyanate, Sigma-Aldrich®
Catalog # SIAL128406-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:153.18 g/mol
Boiling Pt:228 °C (1013 hPa)
Melting Pt:26…28 °C
Storage Temperature:Refrigerator
ADR:8,III
:M-Clarity™ quality level = MQ200
EINECS:216-280-4
UN:2923
CAS Number:1544-68-9
Formula:C₇H₄FNS
MDL Number:MFCD00004809