2-Fluorophenyl isothiocyanate, Sigma-Aldrich®
Catalog # SIAL253693-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:153.18 g/mol
Boiling Pt:54…58 °C (1 mmHg)
Melting Pt:24…26 °C
Density:1.24 g/cm³ (20 °C)
EINECS:254-237-1
UN:1760
ADR:8,II
:M-Clarity™ quality level = MQ100
CAS Number:38985-64-7
Formula:C₇H₄FNS
MDL Number:MFCD00004800