Order Entry
Poland
ContactUsLinkComponent
2,6-Difluorophenyl isothiocyanate, Sigma-Aldrich®
2,6-Difluorophenyl isothiocyanate, Sigma-Aldrich®
Catalog # SIAL511781-5G
Supplier:  Merck
CAS Number:  
2,6-Difluorophenyl isothiocyanate, Sigma-Aldrich®
Catalog # SIAL511781-5G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :171.17 g/mol
Boiling Pt:106…107 °C (35 mmHg)
Storage Temperature:Refrigerator
ADR:6.1,II
 :M-Clarity™ quality level = MQ100
UN:2930
CAS Number:207974-17-2
Formula:C₇H₃F₂NS
MDL Number:MFCD00041047