
2,3,4,5-Tetrafluorobenzoic acid, Sigma-Aldrich®
Catalog # SIAL326267-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:194.09 g/mol
Boiling Pt:135…137 °C (23 mmHg)
Melting Pt:92…92.5 °C
:M-Clarity™ quality level = MQ100
:GHS07
Storage Temperature:Ambient
CAS Number:1201-31-6
Formula:C₇H₂F₄O₂
MDL Number:MFCD00009613