Terephthalic acid 99+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:100-21-0
:
Formula:C₈H₆O₄
MDL Number:MFCD00002558
:166.13 g/mol
Merck Index:12,09306
Boiling Pt:392 °C (1013 hPa)
Melting Pt:>300 °C
Density:1,472 g/cm³ (20 °C)
Flash Pt:260 °C
Storage Temperature:Ambient
EINECS:202-830-0
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Water:≤0.2 %
- Sulfated ash:≤0.001 %
- Titration with TBAH:≥99.0 %
Specification Test Results
Frequently Bought Together