p-Anisic acid, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:100-09-4
:
Formula:C₈H₈O₃
MDL Number:MFCD00002542
:152.15 g/mol
Merck Index:13,00669
Boiling Pt:275 °C (1013 hPa)
Melting Pt:182…185 °C
Density:∼1.385 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ300
Storage Temperature:Ambient
EINECS:202-818-5
Frequently Bought Together