4-hydroksybenzoesan metylu, Sigma-Aldrich®
Supplier: Merck
Jump to:
About this item
Synonyms: Methylparaben, 4-Hydroxybenzoic acid methyl ester, Methyl Parahydroxybenzoate, Methyl 4-hydroxybenzoate
- CAS Number:99-76-3
- Formula:HOC₆H₄CO₂CH₃
- MDL Number:MFCD00002352
- Molecular Weight:152.15 g/mol
- Merck Index:13,06129
- Boiling Pt:270…280 °C (1013 hPa)
- Melting Pt:125…128 °C
- Density:1,1208 g/cm³ (137 °C)
- :M-Clarity™ quality level = MQ200
- Storage Temperature:Temperatura otoczenia
- EINECS:202-785-7
2 Options Available Below
Product Details & Documents
Synonyms: Methylparaben, 4-Hydroxybenzoic acid methyl ester, Methyl Parahydroxybenzoate, Methyl 4-hydroxybenzoate
- CAS Number:99-76-3
- Formula:HOC₆H₄CO₂CH₃
- MDL Number:MFCD00002352
- Molecular Weight:152.15 g/mol
- Merck Index:13,06129
- Boiling Pt:270…280 °C (1013 hPa)
- Melting Pt:125…128 °C
- Density:1,1208 g/cm³ (137 °C)
- :M-Clarity™ quality level = MQ200
- Storage Temperature:Temperatura otoczenia
- EINECS:202-785-7
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



