Order Entry
Poland
Orders LinkContactUsLinkComponent
Methyl-4-hydroxybenzoate

Methyl-4-hydroxybenzoate

Glass bottle for solids, 100 g
Catalog # MOLE14385952-100G
Irritant
Marketsource
Supplier:   Molekula
Methyl-4-hydroxybenzoate
Catalog # MOLE14385952-100G
Supplier:  Molekula
Marketsource
CAS Number:  99-76-3
Methyl-4-hydroxybenzoate
Methyl-4-hydroxybenzoate

Glass bottle for solids, 100 g

CAS Number:  99-76-3
Price
545 zł
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  Methylparaben, 4-Hydroxybenzoic acid methyl ester, Methyl Parahydroxybenzoate, Methyl 4-hydroxybenzoate
  • CAS Number:99-76-3
  • Formula:HOC₆H₄CO₂CH₃
  • MDL Number:MFCD00002352
  • Molecular Weight:152.15 g/mol
  • Merck Index:13,06129
  • Boiling Pt:270…280 °C (1013 hPa)
  • Melting Pt:125…128 °C
  • Density:1.1208 g/cm³ (137 °C)
  • Storage Temperature:Ambient
  • EINECS:202-785-7
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.