Order Entry
Poland
ContactUsLinkComponent
Benzoyl chloride for synthesis, Sigma-Aldrich®
Benzoyl chloride for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Benzoyl chloride for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Solubility:
    (20 °C) (decomposition)
  • Melting Point:
    -0.6 °C
  • Molar Mass:
    140.57 g/mol
  • Boiling Point:
    197.2 °C (1013 hPa)
  • Vapor Pressure:
    0.5 hPa (20 °C)
  • Flash Point:
    72 °C
  • Refractive Index:
    1.5537 (20 °C, 589 nm)
  • Explosion Limit:
    2.5 - 27 %(V)
  • Density:
    1.21 g/cm3 (20 °C)
  • pH:
    2 (1 g/l, H2O, 20 °C)
  • Ignition Point:
    600 °C
  • Hazard Class:
    8
  • Hazard Class Description:
    Corrosive Material

Specification Test Results

Related Information

Frequently Bought Together