

2-Cyanobenzenesulphonyl chloride ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:69360-26-5
:
Formula:C₇H₄ClNO₂S
MDL Number:MFCD00052341
Melting Pt:65…69 °C
Storage Temperature:Ambient
UN:3261
ADR:8,III
:201.63 g/mol
:GHS05
:GHS07
Related Information
Frequently Bought Together