Order Entry
Poland
ContactUsLinkComponent
3,4-Dichlorobenzenesulphonyl chloride ≥98%
Corrosive
3,4-Dichlorobenzenesulphonyl chloride ≥98%
Catalog # A14576.06
CAS Number:  
Corrosive
3,4-Dichlorobenzenesulphonyl chloride ≥98%
Catalog # A14576.06
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications

About this item

 :
 :245.51 g/mol
Boiling Pt:253 °C
Melting Pt:20…22 °C
Density:1.572
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
EINECS:202-656-5
UN:3265
ADR:8,II
 :GHS05
CAS Number:98-31-7
Formula:C₆H₃Cl₃O₂S
MDL Number:MFCD00041255