Order Entry
Poland
ContactUsLinkComponent
 
2,6-Dichlorobenzenesulphonyl chloride, Sigma-Aldrich®
Catalog # SIAL545708-5G
Supplier:  Merck
CAS Number:  
2,6-Dichlorobenzenesulphonyl chloride, Sigma-Aldrich®
Catalog # SIAL545708-5G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
Melting Pt:54…56 °C
UN:3261
ADR:8,II
 :M-Clarity™ quality level = MQ100
 :245.51 g/mol
CAS Number:6579-54-0
Formula:C₆H₃Cl₃O₂S
MDL Number:MFCD00052311