Order Entry
Poland
ContactUsLinkComponent
2,4-Dichlorobenzenesulphonyl chloride, Sigma-Aldrich®
2,4-Dichlorobenzenesulphonyl chloride, Sigma-Aldrich®
Catalog # SIAL545694-5G
Supplier:  Merck
CAS Number:  
2,4-Dichlorobenzenesulphonyl chloride, Sigma-Aldrich®
Catalog # SIAL545694-5G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :245.51 g/mol
Melting Pt:56 °C
ADR:8,II
 :M-Clarity™ quality level = MQ200
UN:3261
CAS Number:16271-33-3
Formula:C₆H₃Cl₃O₂S
MDL Number:MFCD00052712