



3,5-Dichlorophenyl isocyanate ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:34893-92-0
:
Formula:C₇H₃Cl₂NO
MDL Number:MFCD00013859
:188.01 g/mol
Boiling Pt:242…244 °C
Melting Pt:28…32 °C
Density:1.380
Flash Pt:134 °C (273 °F)
EINECS:252-276-9
UN:2250
ADR:6.1,II
:GHS06
:GHS08
:GHS05
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together