Salicylaldehyde, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:90-02-8
:
Formula:C₇H₆O₂
MDL Number:MFCD00003317
:122.12 g/mol
Merck Index:13,08405
Boiling Pt:196 °C (1013 hPa)
Melting Pt:–7 °C
Density:1.17 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Flash Pt:79 °C
EINECS:201-961-0
:GHS07
Storage Temperature:Ambient
Frequently Bought Together