Order Entry
Poland
Orders LinkContactUsLinkComponent
Vanillin

Vanillin

Irritant
Supplier:   Merck
Vanillin
Supplier:  Merck
Certificates
Vanillin

3 Options Available Below

Product Details & Documents
Synonyms:  4-Hydroxy-3-methoxybenzaldehyde
  • CAS Number:121-33-5
  • Formula:C₈H₈O₃
  • MDL Number:MFCD00006942
  • Molecular Weight:152.15 g/mol
  • Merck Index:13,09998
  • Boiling Pt:285 °C (1013 hPa)
  • Melting Pt:∼82 °C
  • Density:1.056 g/cm³ (20 °C)
  •  :M-Clarity™ quality level = MQ300
  • Flash Pt:153 °C
  • Storage Temperature:Ambient
  • EINECS:204-465-2
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 1 110 zł
    1 110 zł
    1 * 100 g
       1 110 zł

  • Glass bottle for solids
    1 kg
    Log in for availability
    Log in for availability
    1 * 1 kg
    1 * 1 kg - 7 300 zł
    7 300 zł
    1 * 1 kg
       7 300 zł

  • 5 kg
    Log in for availability
    Log in for availability
    1 * 5 kg
    1 * 5 kg