Benzaldehyde, Sigma-Aldrich®
Supplier: Merck
Jump to:
About this item
- CAS Number:100-52-7
- Formula:C₆H₅CHO
- MDL Number:MFCD00003299
- Molecular Weight:106.12 g/mol
- Merck Index:13,01057
- Boiling Pt:179 °C (1013 hPa)
- Melting Pt:–26 °C
- Density:1.049 g/cm³ (20 °C)
- Flash Pt:64 °C
- Storage Temperature:Ambient
- UN:1990
- ADR:9,III
- :M-Clarity™ quality level = MQ300
- EINECS:202-860-4
4 Options Available Below
Product Details & Documents
- CAS Number:100-52-7
- Formula:C₆H₅CHO
- MDL Number:MFCD00003299
- Molecular Weight:106.12 g/mol
- Merck Index:13,01057
- Boiling Pt:179 °C (1013 hPa)
- Melting Pt:–26 °C
- Density:1.049 g/cm³ (20 °C)
- Flash Pt:64 °C
- Storage Temperature:Ambient
- UN:1990
- ADR:9,III
- :M-Clarity™ quality level = MQ300
- EINECS:202-860-4
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



