
Benzaldehyde, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:100-52-7
:
Formula:C₆H₅CHO
MDL Number:MFCD00003299
:106.12 g/mol
Merck Index:13,01057
Boiling Pt:179 °C (1013 hPa)
Melting Pt:–26 °C
Density:1.049 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:202-860-4
UN:1990
ADR:9,III
:M-Clarity™ quality level = MQ200
Flash Pt:64 °C
:GHS07
Frequently Bought Together