2-Bromophenyl isothiocyanate, Sigma-Aldrich®
Catalog # SIAL253154-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:214.09 g/mol
Boiling Pt:257 °C (1013 hPa)
Melting Pt:20 °C
Density:1.59 g/cm³ (20 °C)
UN:3334
ADR:9,
:M-Clarity™ quality level = MQ100
CAS Number:13037-60-0
Formula:C₇H₄BrNS
MDL Number:MFCD00004799