Order Entry
Poland
ContactUsLinkComponent
1-Chloro-3,5-dibromobenzene ≥98%
1-Chloro-3,5-dibromobenzene ≥98%
Catalog # B25411.14
CAS Number:  
1-Chloro-3,5-dibromobenzene ≥98%
Catalog # B25411.14
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308
  • Environmentally Preferable:

Specifications

About this item

Synonyms: 3,5-Dibromochlorobenzene, 1,3-Dibromo-5-chlorobenzene
CAS Number:14862-52-3
 :
Formula:C₆H₃Br₂Cl
MDL Number:MFCD00070765
 :270.35 g/mol
 :GHS07
Melting Pt:95…98 °C
Storage Temperature:Ambient