1,4-Dibromobenzene, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:106-37-6
:
Formula:C₆H₄Br₂
MDL Number:MFCD00000089
Boiling Pt:219
Melting Pt:84…87 °C
Density:2.28 g/cm³ (20 °C)
Molecular Weight:235.91 g/mol
Flash Pt:>100 °C
Storage Temperature:Ambient
EINECS:203-390-2
UN:3077
ADR:9 III
Merck Index:13,03042
:M-Clarity™ quality level = MQ100
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together