Cycloheximide, Sigma-Aldrich®
Supplier: Merck
Jump to:
About this item
Synonyms: Naramycin A, Actidione, 3-[2-(3,5-Dimethyl-2-oxocyclohexyl)-2-hydroxyethyl]glutarimide,
- CAS Number:66-81-9
- Formula:C₁₅H₂₃NO₄
- MDL Number:MFCD00082346
- Molecular Weight:281.35 g/mol
- Storage Temperature:Ambient
- ADR:6.1,I
- :M-Clarity™ quality level = MQ200
- EINECS:200-636-0
- UN:2811
2 Options Available Below
Product Details & Documents
Synonyms: Naramycin A, Actidione, 3-[2-(3,5-Dimethyl-2-oxocyclohexyl)-2-hydroxyethyl]glutarimide,
- CAS Number:66-81-9
- Formula:C₁₅H₂₃NO₄
- MDL Number:MFCD00082346
- Molecular Weight:281.35 g/mol
- Storage Temperature:Ambient
- ADR:6.1,I
- :M-Clarity™ quality level = MQ200
- EINECS:200-636-0
- UN:2811



