Succinimide 98%
:123-56-8
:
:C₄H₅NO₂
:MFCD00005495
:99.09 g/mol
:12,09040
:285…290 °C (760 mmHg)
:121…126 °C
:1,418 g/cm³ (20 °C)
:201 °C
:204-635-6
:GHS07
:Ambient
- Appearance:Off-white to beige to light brown Powder or flakes
- Infrared spectrum:Conforms
- Melting point:121 to 126 °C
- Titration with NaOH:≥97.5 %
Frequently Bought Together