Order Entry
Poland
ContactUsLinkComponent
10-Undecin-1-ol, Sigma-Aldrich®
10-Undecin-1-ol, Sigma-Aldrich®
Catalog # SIAL94195-1ML
Supplier:  Merck
CAS Number:  
10-Undecin-1-ol, Sigma-Aldrich®
Catalog # SIAL94195-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • Seq:
    2070

Specifications

About this item

 :
 :168.28 g/mol
CAS Number:2774-84-7
Formula:C₁₁H₂₀O
MDL Number:MFCD00041675