Order Entry
Poland
Orders LinkContactUsLinkComponent
3-Hydroxycinnamic acid

3-Hydroxycinnamic acid

Glass bottle for solids, 100 g
Catalog # MOLE21455098-100G
Irritant
Marketsource
Supplier:   Molekula
3-Hydroxycinnamic acid
Catalog # MOLE21455098-100G
Supplier:  Molekula
Marketsource
CAS Number:  588-30-7
3-Hydroxycinnamic acid

Glass bottle for solids, 100 g

CAS Number:  588-30-7
Price
Inquire For Price
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  m-Hydroxycinnamic acid, 3-(3-Hydroxyphenyl)acrylic acid, m-Coumaric acid
  • CAS Number:588-30-7
  • Formula:C₉H₈O₃
  • MDL Number:MFCD00004390
  • Molecular Weight:164.158 g/mol
  • EINECS:209-615-0
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.