
Cinnamic acid for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:140-10-3
Formula:C₉H₈O₂
MDL Number:MFCD00004369
:148.15 g/mol
:M-Clarity™ quality level = MQ200
EINECS:205-398-1
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Solubility:0.4 g/l (20°C)
- Melting Point:132 - 136 °C
- Molar Mass:500 kg/m3
- Bulk Density:500 kg/m3
- Boiling Point:300 °C (1013 hPa)
- Vapor Pressure:1.3 hPa (128 °C)
- Flash Point:160 °C
- Density:1.25 g/cm3 (4 °C)
- pH:3 - 4 (0.4 g/l, H2O, 20 °C)
- Hill Formula:C9H8O2
- Chemical Formula:C6H5CH=CHCOOH
Specification Test Results
Related Information
Frequently Bought Together