2-Methyl-1-butene, Sigma-Aldrich®
SIAL257486-25G
: Merck
:
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
:
:70.13 g/mol
:31 °C (1013 hPa)
:–137.5 °C
:0.6496 g/cm³ (20 °C)
:Refrigerator
:209-250-7
:2459
:3,I
:M-Clarity™ quality level = MQ100
:563-46-2
:C₅H₁₀
:MFCD00009333