

1-Bromo-3-methyl-2-butene 96%
:870-63-3
:
:C₅H₉Br
:MFCD00000242
:82…83 °C (150 mmHg)
:1.27 g/cm³
:32 °C
:212-799-5
:2920
:8,II
:149.03 g/mol
:GHS02
:GHS05
:Refrigerator
- Appearance:Clear colorless to yellow to brown Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4890 to 1.4910 (20°C, 589 nm)
- Titration Argentometric:≥95 % (ex Br)
- Stabilizer:silver wool
Frequently Bought Together