Order Entry
Poland
ContactUsLinkComponent
cis-3-Hexen-1-ol, Sigma-Aldrich®
cis-3-Hexen-1-ol, Sigma-Aldrich®
Catalog # SIALH12900-10G
Supplier:  Merck
CAS Number:  
cis-3-Hexen-1-ol, Sigma-Aldrich®
Catalog # SIALH12900-10G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

 :
 :100.16 g/mol
Merck Index:13,04719
Boiling Pt:156 °C (1013 hPa)
Melting Pt:–60 °C
Density:0.85 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:213-192-8
UN:1987
ADR:3,III
 :M-Clarity™ quality level = MQ100
CAS Number:928-96-1
Formula:C₆H₁₂O
MDL Number:MFCD00063217