
cis-6-Nonenal, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:2277-19-2
:
Formula:C₉H₁₆O
MDL Number:MFCD00014683
:140.23 g/mol
Boiling Pt:87 °C (1013 hPa)
Density:0.84 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ300
EINECS:218-900-9
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together