
cis-4-Decenal, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:21662-09-9
:
Formula:C₁₀H₁₈O
MDL Number:MFCD00007024
Boiling Pt:78 °C (1013 hPa)
Density:0.8569 g/cm³ (20 °C)
ADR:9,
:M-Clarity™ quality level = MQ300
:154.25 g/mol
EINECS:244-514-5
UN:3334
:GHS07
Frequently Bought Together