Order Entry
Poland
ContactUsLinkComponent
cis-11-Hexadecenal, Sigma-Aldrich®
Irritant
cis-11-Hexadecenal, Sigma-Aldrich®
Catalog # SIAL249084-100MG
Supplier:  Merck
CAS Number:  
Irritant
cis-11-Hexadecenal, Sigma-Aldrich®
Catalog # SIAL249084-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :238.41 g/mol
 :M-Clarity™ quality level = MQ100
EINECS:258-876-7
 :GHS07
Storage Temperature:Refrigerator
CAS Number:53939-28-9
Formula:C₁₆H₃₀O
MDL Number:MFCD00010124