4-(Indol-3-yl)butyric acid, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:133-32-4
:
Formula:C₁₂H₁₃NO₂
MDL Number:MFCD00005664
:203.24 g/mol
Merck Index:13,04987
Melting Pt:121…126 °C
Density:1.27 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:205-101-5
ADR:6.1,III
:M-Clarity™ quality level = MQ200
UN:2811
Frequently Bought Together