
2,5-Dimethyl-2-hexanol ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3730-60-7
:
Formula:C₈H₁₈O
MDL Number:MFCD00021814
:130.23 g/mol
Boiling Pt:155…156 °C
Density:0.8203 g/cm³ (0 °C)
Flash Pt:52 °C (125 °F)
Storage Temperature:Ambient
EINECS:223-085-8
UN:1987
ADR:3,III
:GHS02
Related Information
Frequently Bought Together