Plant cell culture media, Hoagland’s No.2 basal salt mixture
Supplier: Merck
Certificates
About this item
Hoagland’s No.2 basal salt mixture is a classic plant medium with the macro- and micronutrients as described by Hoagland and Arnon (1938).
Quantity: Formulated to contain 1,6 grams of powder per litre of medium.
Application: Hoagland’s solution is a popular solution for a wide variety of plant species.
Specifications
- M-Clarity quality level:MQ200
- Storage temperature:2...8 °C
Specifications
Frequently Bought Together