

Isopropyl isothiocyanate ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2253-73-8
:
Formula:C₄H₇NS
MDL Number:MFCD00037830
:101.17 g/mol
Boiling Pt:137…138 °C
Density:0.948
Flash Pt:40 °C (104 °F)
Storage Temperature:Ambient
EINECS:218-851-3
UN:1993
ADR:3,III
:GHS02
:GHS07
Related Information
Frequently Bought Together