tert-Butyl isothiocyanate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:590-42-1
:
Formula:C₅H₉NS
MDL Number:MFCD00004816
:115.2 g/mol
Boiling Pt:140…143 °C (1013 hPa)
Melting Pt:10.5 °C
Density:0.9187 g/cm³ (10 °C)
EINECS:209-682-6
ADR:6.1,II
:M-Clarity™ quality level = MQ100
UN:2929
Frequently Bought Together