
2-Bromo-2-methylpropane, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:507-19-7
:
Formula:C₄H₉Br
MDL Number:MFCD00000125
:137.02 g/mol
Merck Index:13,01554
Boiling Pt:73 °C (1013 hPa)
Melting Pt:–20 °C
Density:1.2125 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:208-065-9
UN:2342
ADR:3,II
:M-Clarity™ quality level = MQ200
Flash Pt:16 °C
:GHS02
Frequently Bought Together