
1-Bromo-2-methylpropane, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:78-77-3
:
Formula:C₄H₉Br
MDL Number:MFCD00000217
:137.02 g/mol
Merck Index:13,05152
Boiling Pt:91 °C (1013 hPa)
Melting Pt:–118 °C
Density:1.265 g/cm³ (20 °C)
UN:2342
ADR:3,II
:M-Clarity™ quality level = MQ200
EINECS:201-141-2
:GHS02
Storage Temperature:Ambient
Frequently Bought Together