Sodium dodecyl sulphate (SDS), Sigma-Aldrich®
: Merck
:151-21-3
:
:CH₃(CH₂)₁₁OSO₃Na
:MFCD00036175
:288.38 g/mol
:14,08636
:204…207 °C
:1.1 g/cm³ (20 °C)
:>100 °C
:1325
:4.1,II
:M-Clarity™ quality level = MQ200
:205-788-1
:GHS02
:GHS06
:Ambient
Frequently Bought Together