
Aluminium isopropoxide, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:555-31-7
:
Formula:C₉H₂₁AlO₃
MDL Number:MFCD00008870
:204.25 g/mol
Merck Index:13,00346
Boiling Pt:135 °C (13 hPa)
Melting Pt:119 °C
Density:0.9771 g/cm³ (20 °C)
UN:3181
ADR:4.1,II
:M-Clarity™ quality level = MQ200
EINECS:209-090-8
:GHS02
Frequently Bought Together