Order Entry
Poland
ContactUsLinkComponent
Methanol ≥99.9%, HPLC Plus for HPLC, for GC (residue analysis), Sigma-Aldrich®
Flammable
Toxic
Health
Methanol ≥99.9%, HPLC Plus for HPLC, for GC (residue analysis), Sigma-Aldrich®
Catalog # SIAL646377-1L
Supplier:  Merck
New Product
CAS Number:  
Flammable
Toxic
Health
Methanol ≥99.9%, HPLC Plus for HPLC, for GC (residue analysis), Sigma-Aldrich®
Catalog # SIAL646377-1L
Supplier:  Merck
New Product
CAS Number:  

Specifications

  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Appearance (Color):
    Colorless
  • Appearance (Form):
    Liquid
  • Infrared Spectrum:
    Conforms to Structure
  • Color Test:
    < 10 APHA
  • UV Absorbance 400nm:
    < 0.005
  • UV Absorbance 250nm:
    < 0.02
  • UV Absorbance 230nm:
    < 0.10
  • UV Absorbance 220nm:
    < 0.20
  • UV Absorbance 210nm:
    < 0.80
  • UV Absorbance 205nm:
    < 1.0
  • Purity (GC):
    99.90 %
  • Water (by Karl Fischer):
    < 0.05 %
  • Residue on Evaporation:
    < 0.0001 %
  • Fluorescence 365nm as Quinine:
    < 1.0 ppb
  • Fluorescence 254nm:
    < 1.0 ppb
  • Titratable Acid (meq/g):
    < 0.0003
  • Titratable Base (meq/g):
    < 0.0002
  • Carbonyl 0.001% (maximum) each of acetone, formaldehyde, and acetaldehyde:
    Pass
  • HPLC Absorbance 230 nm:
    < 2 mAU
  • HPLC Absorbance 254 nm:
    < 5 mAU
  • Recommended Retest Period:
    5 Years

Specification Test Results

About this item

 :
 :32.04 g/mol
Merck Index:13,05984
Boiling Pt:64.6 °C (1013 hPa)
Melting Pt:–98 °C
Density:0.7918 g/cm³ (20 °C)
Flash Pt:11 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-659-6
UN:1230
ADR:3,II
REACH:01-2119433307-44
 :GHS02
 :GHS06
 :GHS08
CAS Number:67-56-1
Formula:CH₃OH
MDL Number:MFCD00004595