Order Entry
Poland
ContactUsLinkComponent
Glutaric anhydride | Glutaric anhydride
Glutaric anhydride
Catalog # MOLE20533442-500G
Supplier:  Molekula
CAS Number:  
Glutaric anhydride
Catalog # MOLE20533442-500G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

About this item

 :
 :114.1 g/mol
Boiling Pt:287 °C (1013 hPa)
Melting Pt:52…55 °C
Density:1,411 g/cm³ (20 °C)
Flash Pt:174 °C
Storage Temperature:Ambient
EINECS:203-593-6
CAS Number:108-55-4
Formula:C₅H₆O₃
MDL Number:MFCD00006679