Order Entry
Poland
ContactUsLinkComponent
Dibutyl ether for synthesis, Sigma-Aldrich®
Flammable
Irritant
Dibutyl ether for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Flammable
Irritant
Dibutyl ether for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    10 g/l (20 °C)
  • Melting Point:
    -95 °C
  • Molar Mass:
    130.23 g/mol
  • Boiling Point:
    140 - 143 °C (1013 hPa)
  • Vapor Pressure:
    6.4 hPa (20 °C)
  • Flash Point:
    37 - 38 °C
  • Refractive Index:
    1.3985
  • Explosion Limit:
    0.9 - 8.5 %(V)
  • Density:
    0.769 g/cm3 (20 °C)
  • pH:
    5.2 (0.2 g/l, H2O, 20 °C)
  • Ignition Point:
    175 °C
  • Water absorption:
    300 mg/kg (20 °C)
  • Hazard Class:
    3
  • Hazard Class Description:
    Flammable Liquid

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together