Order Entry
Poland
Orders LinkContactUsLinkComponent
Taurine

Taurine

Taurine
Certificates

2 Options Available Below

Product Details & Documents
Synonyms:  2-Aminoethanesulphonic acid
  • CAS Number:107-35-7
  • Formula:C₂H₇NO₃S
  • MDL Number:MFCD00008197
  • Molecular Weight:125.15 g/mol
  • Merck Index:13,09163
  • Melting Pt:300 °C
  • Storage Temperature:Ambient
  • EINECS:203-483-8
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g

  • Glass bottle for solids
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g