Order Entry
Poland
ContactUsLinkComponent
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) ≥99.3% suitable for buffer solutions
Irritant
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) ≥99.3% suitable for buffer solutions
Supplier:  PanReac AppliChem
Certificates
undefined
Irritant
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) ≥99.3% suitable for buffer solutions
Supplier:  PanReac AppliChem

Specification Test Results

  • Assay (titr.):
    min. 99.3 %
  • pH (1 M; H2O; 20°C):
    10.5 - 12.0
  • Heavy metals (as Pb):
    max. 0.001 %
  • Water:
    max. 0.5 %

Specification Test Results

Related Information

Frequently Bought Together