Ethyl isothiocyanate, Sigma-Aldrich®
Catalog # SIALE33904-10G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:87.15 g/mol
Merck Index:13,03848
Boiling Pt:130…132 °C (1013 hPa)
Melting Pt:–6 °C
Density:1 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:208-831-2
UN:2929
ADR:6.1,II
:M-Clarity™ quality level = MQ100
CAS Number:542-85-8
Formula:C₃H₅NS
MDL Number:MFCD00004820