


N,N-Dimethylethylendiamine 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:108-00-9
:
Formula:C₄H₁₂N₂
MDL Number:MFCD00008175
:88.15 g/mol
Boiling Pt:107 °C
Melting Pt:< –70 °C
Density:0.810
Flash Pt:12 °C (53 °F)
EINECS:203-541-2
ADR:3,II
:GHS02
:GHS05
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together